Hydrochlorothiazid
Lieferant: LGC Standards PROMOCHEM
BP186
PROOBP186EA
297
EUR
PROOBP186
Hydrochlorothiazid
Hydrochlorothiazid
Formel:
C₇H₈ClN₃O₄S₂ |
MDL:
MFCD00051765 CAS-Nummer: 58-93-5 EINECS: 200-403-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...