3-(3,4-Dihydroxyphenyl)-L-alanin
Lieferant: LGC Standards PROMOCHEM
Synonyme:
Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa
BP213
PROOBP213EA
298
EUR
PROOBP213
3-(3,4-Dihydroxyphenyl)-L-alanin
3-(3,4-Dihydroxyphenyl)-L-alanin
Formel:
C₉H₁₁NO₄ Molecular Weight: 197,19 g/mol |
MDL:
MFCD00002598 CAS-Nummer: 59-92-7 EINECS: 200-445-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...