Lidocain Hydrochlorid
Lieferant: LGC Standards PROMOCHEM
BP214
PROOBP214EA
298
EUR
PROOBP214
Lidocain Hydrochlorid
Lidocain Hydrochlorid
Formel:
C₁₄H₂₃ClN₂O |
CAS-Nummer:
73-78-9 EINECS: 200-803-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...