1-(2-Methoxyphenyl)piperazin
Lieferant: LGC Standards PROMOCHEM
BP232
PROOBP232EA
255
EUR
PROOBP232
1-(2-Methoxyphenyl)piperazin
1-(2-Methoxyphenyl)piperazin
Formel:
C₁₁H₁₆N₂O Molecular Weight: 192,26 g/mol Siedepunkt: 380 °C (1013 hPa) Schmelzpunkt: 35…38 °C Dichte: 1,09 g/cm³ (20 °C) |
MDL:
MFCD00005958 CAS-Nummer: 35386-24-4 EINECS: 252-537-7 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...