2-Methyl-4(5)-nitroimidazol
Lieferant: LGC Standards PROMOCHEM
Synonyme:
2-Methyl-4-nitroimidazol, 2-Methyl-4-nitro-1H-imidazol
BP244
PROOBP244EA
256
EUR
PROOBP244
2-Methyl-4(5)-nitroimidazol
2-Methyl-4(5)-nitroimidazol
Formel:
C₄H₅N₃O Molecular Weight: 127,10 g/mol |
MDL:
MFCD00005191 CAS-Nummer: 696-23-1 EINECS: 211-790-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...