Monosulfiram

Lieferant: LGC Standards PROMOCHEM

Synonyme: Bis(diethyl-thiocarbamoyl)sulfid, Tetraethylthiuramsulfid, Sulfiram

BP254
PROOBP254EA 256 EUR
PROOBP254
Monosulfiram
Monosulfiram
Formel: C₁₀H₂₀N₂S₃
Molecular Weight: 264,48 g/mol
CAS-Nummer: 95-05-6
EINECS: 202-387-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR