Theophyllin

Lieferant: LGC Standards PROMOCHEM

Synonyme: 1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline

BP327
PROOBP327EA 297 EUR
PROOBP327
Theophyllin
Theophyllin
Formel: C₇H₈N₄O₂
Molecular Weight: 180,16 g/mol
MDL: MFCD00079619
CAS-Nummer: 58-55-9
EINECS: 200-385-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR