3,5,6-Trichlorsalicylsäure
Lieferant: LGC Standards PROMOCHEM
Synonyme:
3,5,6-Trichlorosalicylic Acid
BP341
PROOBP341EA
256
EUR
PROOBP341
3,5,6-Trichlorsalicylsäure
3,5,6-Trichlorsalicylsäure
Formel:
C₇H₃Cl₃O₃ Molecular Weight: 241,46 g/mol |
MDL:
MFCD00075245 CAS-Nummer: 40932-60-3 EINECS: 255-144-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...