5-Chlor-2-(methylamino)benzophenon

Lieferant: LGC Standards PROMOCHEM

Synonyme: (5-Chloro-2-(methylamino)phenyl)(phenyl)methanone

BP378
PROOBP378EA 256 EUR
PROOBP378
5-Chlor-2-(methylamino)benzophenon
5-Chlor-2-(methylamino)benzophenon
Formel: C₁₄H₁₂ClNO
Molecular Weight: 245,71 g/mol
MDL: MFCD00008284
CAS-Nummer: 1022-13-5
EINECS: 213-822-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR