2-(4-Methyl-1,3-thiazol-5-yl)ethanol

Lieferant: LGC Standards PROMOCHEM

Synonyme: 5-(2-Hydroxyethyl)-4-methylthiazol, 4-Methyl-5-(2-hydroxyethyl)thiazol, 4-Methyl-5-thiazoleethanol

BP444
PROOBP444EA 256 EUR
PROOBP444
2-(4-Methyl-1,3-thiazol-5-yl)ethanol
2-(4-Methylthiazol-5-yl)ethanol
Formel: C₆H₉NOS
Molecular Weight: 143,21 g/mol
Siedepunkt: 135 °C (1013 hPa)
Dichte: 1,199 g/cm³ (20 °C)
MDL: MFCD00005339
CAS-Nummer: 137-00-8
EINECS: 205-272-6
Merck Index: 12,06208

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR