4'-Isobutylacetophenon
Lieferant: LGC Standards PROMOCHEM
Synonyme:
1-(4-Isobutylphenyl)ethanon, 4-Isobutyl-1-acetylbenzol, p-Isobutylacetophenon
BP557
PROOBP557EA
256
EUR
PROOBP557
4'-Isobutylacetophenon
4'-Isobutylacetophenon
Formel:
C₁₂H₁₆O Molecular Weight: 176,26 g/mol |
MDL:
MFCD00027393 CAS-Nummer: 38861-78-8 EINECS: 254-159-8 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...