Hydrocortison-21-acetat
Lieferant: LGC Standards PROMOCHEM
Synonyme:
Hydrocortisone acetate
BP584
PROOBP584EA
297
EUR
PROOBP584
Hydrocortison-21-acetat
Hydrocortison-21-acetat
Formel:
C₂₃H₃₂O₆ Molecular Weight: 404,51 g/mol |
MDL:
MFCD00037714 CAS-Nummer: 50-03-3 EINECS: 200-004-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...