Diclofenac-Natrium

Lieferant: LGC Standards PROMOCHEM

BP619
PROOBP619EA 298 EUR
PROOBP619
Diclofenac-Natrium
Diclofenac-Natrium
Formel: C₁₄H₁₀Cl₂NNaO₂
MDL: MFCD00082251
CAS-Nummer: 15307-79-6
EINECS: 239-346-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR