Lidocain

Lieferant: LGC Standards PROMOCHEM

BP727
PROOBP727EA 297 EUR
PROOBP727
Lidocain
Lidocain
Formel: C₁₄H₂₂N₂O
MDL: MFCD00026733
CAS-Nummer: 137-58-6
EINECS: 205-302-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR