n-Nonadecan

Lieferant: LGC Standards PROMOCHEM

Synonyme: N-nonadecane

CHE171
PROOCHE171EA 204 EUR
PROOCHE171
n-Nonadecan
n-Nonadecan
Formel: H₃C(CH₂)₁₇CH₃
Molecular Weight: 268,53 g/mol
Siedepunkt: 329,7 °C (1013 hPa)
Schmelzpunkt: 32,5 °C
Dichte: 0,7845 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009012
CAS-Nummer: 629-92-5
EINECS: 211-116-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR