Octachlornaphthalin

Lieferant: LGC Standards PROMOCHEM
Achtung

CIL-ECN-5280
PROOCIL-ECN-5280EA 965 EUR
PROOCIL-ECN-5280
Octachlornaphthalin
Octachlornaphthalin
Formel: C₁₀Cl₈
Molecular Weight: 403.73 g/mol
CAS-Nummer: 2234-13-1
EINECS: 218-778-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR