2,6-Dichlorbiphenyl (PCB Nr. 10)
Lieferant: LGC Standards PROMOCHEM
CIL-PCB-10-C
PROOCIL-PCB-10-CEA
615
EUR
PROOCIL-PCB-10-C
2,6-Dichlorbiphenyl (PCB Nr. 10)
2,6-Dichlorbiphenyl (PCB Nr. 10)
Formel:
C₁₂H₈Cl₂ Molecular Weight: 223,1 g/mol |
CAS-Nummer:
33146-45-1 UN: 3432 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...