2,2',4,4',6-Pentachlorbiphenyl (PCB Nr. 100)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-100-C
PROOCIL-PCB-100-CEA 677 EUR
PROOCIL-PCB-100-C
2,2',4,4',6-Pentachlorbiphenyl (PCB Nr. 100)
2,2',4,4',6-Pentachlorbiphenyl (PCB Nr. 100)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 39485-83-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR