2,3,3',4,4'-Pentachlorbiphenyl (PCB Nr. 105)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-105-C
PROOCIL-PCB-105-CEA 704 EUR
PROOCIL-PCB-105-C
2,3,3',4,4'-Pentachlorbiphenyl (PCB Nr. 105)
2,3,3',4,4'-Pentachlorbiphenyl (PCB Nr. 105)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 32598-14-4
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR