2,3,3',4',5-Pentachlorbiphenyl (PCB Nr. 107)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-107-C
PROOCIL-PCB-107-CEA 677 EUR
PROOCIL-PCB-107-C
2,3,3',4',5-Pentachlorbiphenyl (PCB Nr. 107)
2,3,3',4',5-Pentachlorbiphenyl (PCB Nr. 107)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 70424-68-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR