2,3,3',4,5'-Pentachlorbiphenyl (PCB Nr. 108)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-108-C
PROOCIL-PCB-108-CEA 677 EUR
PROOCIL-PCB-108-C
2,3,3',4,5'-Pentachlorbiphenyl (PCB Nr. 108)
2,3,3',4,5'-Pentachlorbiphenyl (PCB Nr. 108)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 70362-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR