2,3,3',4,6-Pentachlorbiphenyl (PCB Nr. 109)
Lieferant: LGC Standards PROMOCHEM
CIL-PCB-109-C
PROOCIL-PCB-109-CEA
766
EUR
PROOCIL-PCB-109-C
2,3,3',4,6-Pentachlorbiphenyl (PCB Nr. 109)
2,3,3',4,6-Pentachlorbiphenyl (PCB Nr. 109)
Formel:
C₁₂H₅Cl₅ Molecular Weight: 326.44 g/mol |
CAS-Nummer:
74472-35-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...