3,3'-Dichlorbiphenyl (PCB Nr. 11)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-11-C
PROOCIL-PCB-11-CEA 677 EUR
PROOCIL-PCB-11-C
3,3'-Dichlorbiphenyl (PCB Nr. 11)
3,3'-Dichlorbiphenyl (PCB Nr. 11)
Formel: C₁₂H₈Cl₂
Molecular Weight: 223,1 g/mol
CAS-Nummer: 2050-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR