2,3,3',5,5'-Pentachlorbiphenyl (PCB Nr. 111)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-111-C
PROOCIL-PCB-111-CEA 766 EUR
PROOCIL-PCB-111-C
2,3,3',5,5'-Pentachlorbiphenyl (PCB Nr. 111)
2,3,3',5,5'-Pentachlorbiphenyl (PCB Nr. 111)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 39635-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR