2,3,3',5,6-Pentachlorbiphenyl (PCB Nr. 112)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-112-C
PROOCIL-PCB-112-CEA 677 EUR
PROOCIL-PCB-112-C
2,3,3',5,6-Pentachlorbiphenyl (PCB Nr. 112)
2,3,3',5,6-Pentachlorbiphenyl (PCB Nr. 112)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 74472-36-9
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR