2,3',4,4',5-Pentachlorbiphenyl (PCB Nr. 118)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-118-C
PROOCIL-PCB-118-CEA 695 EUR
PROOCIL-PCB-118-C
2,3',4,4',5-Pentachlorbiphenyl (PCB Nr. 118)
2,3',4,4',5-Pentachlorbiphenyl (PCB Nr. 118)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 31508-00-6
UN: 2315
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR