2,3',4,5,5'-Pentachlorbiphenyl (PCB Nr. 120)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-120-C
PROOCIL-PCB-120-CEA 677 EUR
PROOCIL-PCB-120-C
2,3',4,5,5'-Pentachlorbiphenyl (PCB Nr. 120)
2,3',4,5,5'-Pentachlorbiphenyl (PCB Nr. 120)
Formel: C₁₂H₅Cl₅
CAS-Nummer: 68194-12-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR