2,3,3',4',5'-Pentachlorbiphenyl (PCB Nr. 122)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-122-C
PROOCIL-PCB-122-CEA 677 EUR
PROOCIL-PCB-122-C
2,3,3',4',5'-Pentachlorbiphenyl (PCB Nr. 122)
2,3,3',4',5'-Pentachlorbiphenyl (PCB Nr. 122)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 76842-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR