2,3',4,4',5'-Pentachlorbiphenyl (PCB Nr. 123)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-123-C
PROOCIL-PCB-123-CEA 677 EUR
PROOCIL-PCB-123-C
2,3',4,4',5'-Pentachlorbiphenyl (PCB Nr. 123)
2,3',4,4',5'-Pentachlorbiphenyl (PCB Nr. 123)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 65510-44-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR