3,3',4,4',5-Pentachlorbiphenyl (PCB Nr. 126)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-126-C
PROOCIL-PCB-126-CEA 677 EUR
PROOCIL-PCB-126-C
3,3',4,4',5-Pentachlorbiphenyl (PCB Nr. 126)
3,3',4,4',5-Pentachlorbiphenyl (PCB Nr. 126)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 57465-28-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR