3,4'-Dichlorbiphenyl (PCB Nr. 13)
Lieferant: LGC Standards PROMOCHEM
Synonyme:
3,4'-PCB, 1-Chlor-3-(4-chlorphenyl)benzol, PCB Nr. 13
CIL-PCB-13-C
PROOCIL-PCB-13-CEA
677
EUR
PROOCIL-PCB-13-C
3,4'-Dichlorbiphenyl (PCB Nr. 13)
3,4'-Dichlorbiphenyl (PCB Nr. 13)
Formel:
C₁₂H₈Cl₂ Molecular Weight: 223,1 g/mol |
MDL:
MFCD00152402 CAS-Nummer: 2974-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...