2,2',3,3',4,5'-Hexachlorbiphenyl (PCB Nr. 130)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-130-C
PROOCIL-PCB-130-CEA 677 EUR
PROOCIL-PCB-130-C
2,2',3,3',4,5'-Hexachlorbiphenyl (PCB Nr. 130)
2,2',3,3',4,5'-Hexachlorbiphenyl (PCB Nr. 130)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 52663-66-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR