2,2',3,3',5,5'-Hexachlorbiphenyl (PCB Nr. 133)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-133-C
PROOCIL-PCB-133-CEA 677 EUR
PROOCIL-PCB-133-C
2,2',3,3',5,5'-Hexachlorbiphenyl (PCB Nr. 133)
2,2',3,3',5,5'-Hexachlorbiphenyl (PCB Nr. 133)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 35694-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR