2,2',3,3',5,6-Hexachlorbiphenyl (PCB Nr. 134)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-134-C
PROOCIL-PCB-134-CEA 677 EUR
PROOCIL-PCB-134-C
2,2',3,3',5,6-Hexachlorbiphenyl (PCB Nr. 134)
2,2',3,3',5,6-Hexachlorbiphenyl (PCB Nr. 134)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 52704-70-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR