2,2',3,4',5,5'-Hexachlorbiphenyl (PCB Nr. 146)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-146-C
PROOCIL-PCB-146-CEA 907 EUR
PROOCIL-PCB-146-C
2,2',3,4',5,5'-Hexachlorbiphenyl (PCB Nr. 146)
2,2',3,4',5,5'-Hexachlorbiphenyl (PCB Nr. 146)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 51908-16-8
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR