2,2',3,4',5,6-Hexachlorbiphenyl (PCB Nr. 147)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-147-C
PROOCIL-PCB-147-CEA 677 EUR
PROOCIL-PCB-147-C
2,2',3,4',5,6-Hexachlorbiphenyl (PCB Nr. 147)
2,2',3,4',5,6-Hexachlorbiphenyl (PCB Nr. 147)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 68194-13-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR