2,2',3,4',5',6-Hexachlorbiphenyl (PCB Nr. 149)
Lieferant: LGC Standards PROMOCHEM
CIL-PCB-149-C
PROOCIL-PCB-149-CEA
713
EUR
PROOCIL-PCB-149-C
2,2',3,4',5',6-Hexachlorbiphenyl (PCB Nr. 149)
2,2',3,4',5',6-Hexachlorbiphenyl (PCB Nr. 149)
Formel:
C₁₂H₄Cl₆ Molecular Weight: 360.88 g/mol |
CAS-Nummer:
38380-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...