2,2',3,4',6,6'-Hexachlorbiphenyl (PCB Nr. 150)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-150-C
PROOCIL-PCB-150-CEA 677 EUR
PROOCIL-PCB-150-C
2,2',3,4',6,6'-Hexachlorbiphenyl (PCB Nr. 150)
2,2',3,4',6,6'-Hexachlorbiphenyl (PCB Nr. 150)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 68194-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR