2,2',4,4',5,6'-Hexachlorbiphenyl (PCB Nr. 154)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-154-C
PROOCIL-PCB-154-CEA 668 EUR
PROOCIL-PCB-154-C
2,2',4,4',5,6'-Hexachlorbiphenyl (PCB Nr. 154)
2,2',4,4',5,6'-Hexachlorbiphenyl (PCB Nr. 154)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 60145-22-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR