2,3,3',4,4',5-Hexachlorbiphenyl (PCB Nr. 156)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-156-C
PROOCIL-PCB-156-CEA 695 EUR
PROOCIL-PCB-156-C
2,3,3',4,4',5-Hexachlorbiphenyl (PCB Nr. 156)
2,3,3',4,4',5-Hexachlorbiphenyl (PCB Nr. 156)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 38380-08-4
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR