2,3,3',4,4',6-Hexachlorbiphenyl (PCB Nr. 158)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-158-C
PROOCIL-PCB-158-CEA 677 EUR
PROOCIL-PCB-158-C
2,3,3',4,4',6-Hexachlorbiphenyl (PCB Nr. 158)
2,3,3',4,4',6-Hexachlorbiphenyl (PCB Nr. 158)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 74472-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR