2,2',3-Trichlorbiphenyl (PCB Nr. 16)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-16-C
PROOCIL-PCB-16-CEA 677 EUR
PROOCIL-PCB-16-C
2,2',3-Trichlorbiphenyl (PCB Nr. 16)
2,2',3-Trichlorbiphenyl (PCB Nr. 16)
Formel: C₁₂H₇Cl₃
Molecular Weight: 257.55 g/mol
CAS-Nummer: 38444-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR