2,3,3',4',5,5'-Hexachlorbiphenyl (PCB Nr. 162)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-162-C
PROOCIL-PCB-162-CEA 713 EUR
PROOCIL-PCB-162-C
2,3,3',4',5,5'-Hexachlorbiphenyl (PCB Nr. 162)
2,3,3',4',5,5'-Hexachlorbiphenyl (PCB Nr. 162)
Formel: C₁₂H₄Cl₆
CAS-Nummer: 39635-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR