2,3,3',4',5,6-Hexachlorbiphenyl (PCB Nr. 163)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-163-C
PROOCIL-PCB-163-CEA 677 EUR
PROOCIL-PCB-163-C
2,3,3',4',5,6-Hexachlorbiphenyl (PCB Nr. 163)
2,3,3',4',5,6-Hexachlorbiphenyl (PCB Nr. 163)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 74472-44-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR