2,3,3',5,5',6-Hexachlorbiphenyl (PCB Nr. 165)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-165-C
PROOCIL-PCB-165-CEA 677 EUR
PROOCIL-PCB-165-C
2,3,3',5,5',6-Hexachlorbiphenyl (PCB Nr. 165)
2,3,3',5,5',6-Hexachlorbiphenyl (PCB Nr. 165)
Formel: C₁₂H₄Cl₆
CAS-Nummer: 74472-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR