2,3',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 167)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-167-C
PROOCIL-PCB-167-CEA 660 EUR
PROOCIL-PCB-167-C
2,3',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 167)
2,3',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 167)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
Siedepunkt: 383
Schmelzpunkt: 125…127 °C
CAS-Nummer: 52663-72-6
UN: 3432
ADR: 9 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR