3,3',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 169)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-169-C
PROOCIL-PCB-169-CEA 660 EUR
PROOCIL-PCB-169-C
3,3',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 169)
3,3',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 169)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
Siedepunkt: 383
Schmelzpunkt: 208…210 °C
CAS-Nummer: 32774-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR