2,2',4-Trichlorbiphenyl (PCB Nr. 17)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-17-C
PROOCIL-PCB-17-CEA 677 EUR
PROOCIL-PCB-17-C
2,2',4-Trichlorbiphenyl (PCB Nr. 17)
2,2',4-Trichlorbiphenyl (PCB Nr. 17)
Formel: C₁₂H₇Cl₃
Molecular Weight: 257.55 g/mol
CAS-Nummer: 37680-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR