2,2',3,3',4,4',5-Heptachlorbiphenyl (PCB Nr. 170)
Lieferant: LGC Standards PROMOCHEM
CIL-PCB-170-C
PROOCIL-PCB-170-CEA
677
EUR
PROOCIL-PCB-170-C
2,2',3,3',4,4',5-Heptachlorbiphenyl (PCB Nr. 170)
2,2',3,3',4,4',5-Heptachlorbiphenyl (PCB Nr. 170)
Formel:
C₁₂H₃Cl₇ Molecular Weight: 395.32 g/mol |
CAS-Nummer:
35065-30-6 UN: 3432 ADR: 9 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...