2,2',3,3',4,4',6-Heptachlorbiphenyl (PCB Nr. 171)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-171-C
PROOCIL-PCB-171-CEA 677 EUR
PROOCIL-PCB-171-C
2,2',3,3',4,4',6-Heptachlorbiphenyl (PCB Nr. 171)
2,2',3,3',4,4',6-Heptachlorbiphenyl (PCB Nr. 171)
Formel: C₁₂H₃Cl₇
Molecular Weight: 395.32 g/mol
CAS-Nummer: 52663-71-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR